Orders LinkContactUsLinkComponent
Sedimentation cones, Imhoff type, DURAN®
Sedimentation cones, Imhoff type, DURAN®
Imhoff cones, scale up to 1000 ml, ring mark at 1000 ml, Without stopcock
Katalogové číslo SCOT214015403
Dodavatel:   DWK Life Sciences
Sedimentation cones, Imhoff type, DURAN®
Katalogové číslo SCOT214015403
Dodavatel:  DWK Life Sciences
Sedimentation cones, Imhoff type, DURAN®
Sedimentation cones, Imhoff type, DURAN®
Imhoff cones, scale up to 1000 ml, ring mark at 1000 ml, Without stopcock
Přejít na:
Price
3 180,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • Very good chemical resistance
  • Without stopcock
The conical sedimentation cone with graduation according to Imhoff allows a quantification of sediments from a suspension (e.g. in waste water).
Certifications: According to DIN 12672.
Specifikace
  • Type:
    Without stopcock
  • Popis:
    Imhoff cones, scale up to 1000 ml, ring mark at 1000 ml