Sample tubes, with screw caps
15 ml, 110 mm, 25 mm
Catalog # KART1671
Supplier: KARTELL
Jump to:
Product Details & Documents
PFA, translucent.
- Very good chemical resistance
- Very good temperature resistance
With screw cap.
Specifications
- Ø vněj.:25 mm
- Kapacita:15 ml
- Délka:110 mm