Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Hydroxyethyl-methakrylát
2-Hydroxyethyl-methakrylát, Sigma-Aldrich®
2-Hydroxyethyl-methakrylát, Sigma-Aldrich®
2-Hydroxyethyl-methakrylát, Sigma-Aldrich®
SIAL17348-250ML-F
:
Merck
:
2-Hydroxyethyl-methakrylát, Sigma-Aldrich®
SIAL17348-250ML-F
:
Merck
:
Typ balení:
Glass bottle
Bal.:
250 mL
Sekvence:
2590
:
:
212-782-2
:
GHS07
:
868-77-9
:
C₆H₁₀O₃
:
MFCD00002863