Snap cap for round bottom tubes, Falcon®
734-1303
: Corning
- Pro zkumavky:12×75 mm
- Popis:Snap cap, dual position, sterile, LDPE
- Environmentally Preferable:Low Manufacturing Impact
- Certifikát ochrany životního prostředí:I-RECs, ISO 14001 - Reynosa
- Bal.:2.000
Falcon® snap caps are designed for use with 12×75 mm test tubes
These polyethylene caps feature a patented, two-position fit, loose for aerobic culturing, tight for sealing. Sterile.
: Snap caps are packed 500 per pack, 2000 per case.