Combined pH Electrodes
O tomto produktu
Combined glass pH electrodes for direct pH measurement and acid/base titrations in aqueous or non-aqueous media. The all-rounder for a large variety of uncritical applications in almost all kinds of industries: Acid content of beverages, acid and base capacity of water samples, pH start applications.
- Integrated glass sleeve for secure fixation in the titration head
- With S7 screw cap and detachable cap or with Plug & Play S12 connection
- Plug & Play sensor features an integrated sensor chip which allows data storing and monitoring
DG111-SC/DGi111-SC/DG114-SC/DGi114-SC: ARGENTHAL™ reference system
DG113-SC/DGi113-SC: Ag/AgCl/LiCl reference system
DGi101-SC/DG115-SC/DGi115-SC: ARGENTHAL™ reference system with silver ion trap
DGi116-Solvent: ARGENTHAL™/LiCl reference system
Ordering information: Each electrode is supplied with 25 ml of reference electrolyte, individual quality certificate and unique serial number.
3 Dostupných variant níže
Product Details & Documents
Combined glass pH electrodes for direct pH measurement and acid/base titrations in aqueous or non-aqueous media. The all-rounder for a large variety of uncritical applications in almost all kinds of industries: Acid content of beverages, acid and base capacity of water samples, pH start applications.
- Integrated glass sleeve for secure fixation in the titration head
- With S7 screw cap and detachable cap or with Plug & Play S12 connection
- Plug & Play sensor features an integrated sensor chip which allows data storing and monitoring
DG111-SC/DGi111-SC/DG114-SC/DGi114-SC: ARGENTHAL™ reference system
DG113-SC/DGi113-SC: Ag/AgCl/LiCl reference system
DGi101-SC/DG115-SC/DGi115-SC: ARGENTHAL™ reference system with silver ion trap
DGi116-Solvent: ARGENTHAL™/LiCl reference system
Ordering information: Each electrode is supplied with 25 ml of reference electrolyte, individual quality certificate and unique serial number.
Product Family Options
Informace o produktu
- SekvenceTypepHTepl.MateriálElektrolytDiafragmaØ×DPřipojeníDostupnostPrice
- 662-2478Combined glass pH electrode DGi113-SC, plug & play, with sensor chipCombined glass pH electrode DGi113-SC, plug & play, with sensor chip07DGi113-SC, Plug & Play0 - 120...60 °CSklo1 mol/L LiClMovable PTFE sleeve junction12×149,5 mmS12Přihlásit se pro dostupnost1 * 1 KS19 270,00 Kč
- 662-2475Combined glass pH electrode DGi111-SC, plug & play, with sensor chipCombined glass pH electrode DGi111-SC, plug & play, with sensor chip09DGi111-SC, Plug & Play0 - 140...60 °CSklo3 mol/L KClCeramic frit12×149,5 mmS12Přihlásit se pro dostupnost1 * 1 KS15 820,00 Kč
- 662-1669Combined glass pH electrode DGi115-SC, Plug & Play, with sensor chipCombined glass pH electrode DGi115-SC, Plug & Play, with sensor chip11DGi115-SC, Plug & Play0 - 140...+100 °CSklo3 mol/L KClFixed ground glass sleeve junction12×149,5 mmS12Přihlásit se pro dostupnost1 * 1 KS17 970,00 Kč