Clamps
Dodavatel: Lenz Laborglas
Přejít na:
O tomto produktu
Stainless steel.
- Excellent chemical resistance
- Excellent temperature resistance
For fixing lids onto reaction vessels, available for flat flanges LF100-200.
3 Dostupných variant níže
Product Details & Documents
Stainless steel.
- Excellent chemical resistance
- Excellent temperature resistance
For fixing lids onto reaction vessels, available for flat flanges LF100-200.
Product Family Options
Informace o produktu
- SekvenceUrčeno proDostupnostPrice