Pipette stand, rotary
Katalogové číslo HIRS8902101
Dodavatel: HIRSCHMANN
Specifikace
- Ø×H:470×230 mm
- Určeno pro:94 volumetric pipettes
- Barva:Modrá
- Popis:Pipette stand, rotary
- Materiál:PP
- Bal.:1
Specifikace
O tomto produktu
Pipette stand made from blue PP.
- Rotatable model with a reinforced base and an additional, rotatable disk to protect the pipette tips
- 94 holes with different diameters for graduated and volumetric pipettes of any size