Reservoir bottles for automatic pipette / tilt measures
Catalog # BRND126963
Supplier: Brand
Specifications
- NZ:29/32
- Kapacita:1000 ml
- Popis:Reservoir bottle for bottle top tilt measures
- Bal.:1
Specifications
About this item
Soda-lime glass.
Capacity 1000 ml
Neck size 29/32
Ordering information: Please order dispenser/tilt measure separately.