Burette stopcocks, straight, with PTFE spindle
Catalog # HIRS8901060
Supplier: HIRSCHMANN
Specifications
- Určeno pro:2 - 10 ml burettes
- Sekvence:01
- Bal.:1
Specifications
About this item
DURAN®, borosilicate glass, clear, with PTFE spindle.
- Excellent chemical resistance
- Excellent temperature resistance
Straight for burettes.