Gas inlet tubes with valve stopcock
Catalog # 201-2612
Supplier: Lenz Laborglas GmbH & CO.KG
Specifications
- Rozměry zábrusu:14/23 NS
- Bal.:1
Specifications
About this item
DURAN®, borosilicate glass.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
With valve stopcock for dosing, with glass olive (angle: 90°), with cone NS14/23, tube length from top of the joint: approx. 150 mm.