Distilling bridge for short path distillation
Katalogové číslo LENZ05058001
Dodavatel: Lenz Laborglas GmbH & CO.KG
Specifikace
- Rozměry zábrusu:14/23 NS
- Velikost objímky:14/23 NS
- Bal.:1
Specifikace
O tomto produktu
DURAN®, borosilicate glass.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Short condensate path due to compact design of condenser and vacuum connection, with two cones NS 14/23, one socket NS 14/23 and three DIN threads GL 14.