Membrane filters, track etched
Dodavatel: Sartorius
Certifikáty
O tomto produktu
Membrane filters manufactured from high grade PC film using track etch technology. Applicable for particulate analysis, epifluorescence miroscopy, fluid clarification, cytology, cell biology, bioassays, water microbiology, environmental analysis.
- Precise and uniform capillary pore structure with narrow pore size distribution
- Excellent surface capture and high sample visibility
- Smooth, flat surface membranes result in high particulate visibility
Specifikace
- Catalog No:
- SART23006-25-N
- 611-1162
- SART23007-25-N
- SART23007-50-N
- SART23004-25-N
- 611-1163
- Typ:
- 23006
- 23006
- 23007
- 23007
- 23004
- 23007
- Velikost póru:
- 0,40 µm
- 0,40 µm
- 0,20 µm
- 0,20 µm
- 0,80 µm
- 0,20 µm
- Tloušťka:
- 23 - 27 µm
- 23 - 27 µm
- 23 - 27 µm
- 23 - 27 µm
- 23 - 27 µm
- 23 - 27 µm
- Objemový průtok vody:
- 30 ml/min/cm² (1 bar)
- 30 ml/min/cm² (1 bar)
- 10 ml/min/cm² (1 bar)
- 10 ml/min/cm² (1 bar)
- 40 ml/min/cm² (0,7 bar)
- 10 ml/min/cm² (1 bar)
- Maximální teplota:
- 140 °C
- 140 °C
- 140 °C
- 140 °C
- 140 °C
- 140 °C
Specifikace
Související informace
Frequently Bought Together