Microbiological filtration system, MBS II, Whatman™
Catalog # 514-0054
Supplier: Whatman products (Cytiva)
Specifications
- Type:MBS II
- Popis:NC membrane, 0,45 µm, black, white grid
- Bal.:24
Specifications
About this item
The microbiological filtration system is used for optimal microbiological control using membranes. The design of the system, which consists of an electrical membrane dispenser, a funnel dispenser, and a vacuum manifold, leads to more reproducible results.
- Shorter preparation time
- High reproducibility
- Large funnel capacity for foaming liquids
- Funnels can be autoclaved up to 50 times
The special sealing technique ensures easy handling and a good integrity of the funnel and membrane during filtration. This reduces any cross contamination to a minimum.