Kyselina kapronová, Supelco®
Catalog # 21529-5ML
Supplier: Merck
CAS Number:
Specifications
- Typ balení:Glass bottle
- Bal.:5 mL
- Sekvence:2210
Specifications
About this item
:
Merck Index:13,01765
Boiling Pt:202 °C (1013 hPa)
Density:0,9276 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:2829
ADR:8,III
:M-Clarity™ quality level = MQ100
Flash Pt:102 °C
EINECS:205-550-7
:GHS05
CAS Number:142-62-1
Formula:C₆H₁₂O₂
MDL Number:MFCD00004421