Aceton, EMPLURA®, Reagent Grade for synthesis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:67-64-1
:
Formula:CH₃COCH₃
MDL Number:MFCD00008765
Merck Index:13,00067
Boiling Pt:56,2 °C (1013 hPa)
Density:0,792 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-662-2
UN:1090
ADR:3,II
:M-Clarity™ quality level = MQ200
REACH:01-2119471330-49
:GHS02
:GHS07
Related Information
Frequently Bought Together