Low speed orbital shakers, Corning® LSE™
Catalog # CORN6781-FP
Supplier: Corning
Specifications
- Orbit:19 mm
- Shaking motion:Orbital
- Popis:LSE low speed orbital shaker, includes 30×30 cm platform, analog control, staining and destaining gels, EU-Plug
- Environmentally Preferable:
- Model:LSE™
- Platform W×D:300×300 mm
- Power:230 W
- Speed:3 - 60 min⁻¹ with 19 mm orbit
- Hmotnost:5 kg
- Bal.:1