Vacuum check valve and positive connector, Nalgene®
Katalogové číslo NALG6120-0010
Dodavatel: Thermo Fisher Scientific
Specifikace
- Popis:Vacuum check valve and positive connector, 19,05 mm length
- Materiál:HDPE
- Environmentally Preferable:Low Manufacturing Impact
- Certifikát ochrany životního prostředí:NAR Certificate of Retirement
- Velikost hadice/trubice:6,35 - 7,94 mm ID
- Bal.:6
Specifikace
O tomto produktu
These vacuum check valves with positive connector are designed to prevent back pressure with aspirator-type vacuum pumps.
- Work best between 10- and 28- in. Hg.
- Silicone diaphragm prevents back pressure