Caps for closed dome tanks with mixer support assembly, Nalgene®
Catalog # NALG2651-0200
Supplier: Thermo Fisher Scientific
Specifications
- Velikost víčka:152,3 mm
- Popis:with silicone gasket, true union clamp
- Environmentally Preferable:Low Manufacturing Impact
- Bal.:1
- Pro láhve:75/115/ 210/380 l
- Certifikát ochrany životního prostředí:NAR Certificate of Retirement
Specifications
About this item
Polypropylene, PVDF, silicone.
- 6" (15,2 cm) polypropylene screw closure with 2" (5,1 cm) sanitary ferrule welded in the center
- 2" (5,1 cm) silicone gasket and PDVF True Union clamp
- Autoclavable
Use with Nalgene closed-dome tanks (Series 2650).