A mixture of at least three proteolytic activities including an extracellular serine protease. In general, serine proteases display a wide range of substrate specificities, which are believed to be mediated by an active site composed of one Asp, one His, and a Ser residue in the molecule. This enzyme prefers to hydrolyze peptide bonds on the carboxyl side of glutamic or aspartic acid.
Protease is typically used in nucleic acid isolation procedures in incubations of 0,5 to 3,0 hours supplemented with 0.2% sodium dodecyl sulfate and 10 mM EDTA.This enzyme is more active at a higher pH range than the known alkaline protease, showing the proteolytic activity even in 0.2N NaOH solution. This enzyme is useful for proteolysis of insoluble protein and for structure investigation of protein.
- Čí. CAS:9036-06-0
- Zdroj:Streptomyces griseus
- Podmínky uskladnění:Freezer
- MDL čí.:MFCD02253017
- Název enzymu:Pronase E
- Koncentrace:≥3.5 units/mg solid