Accessories for aspirating pipettes, Costar®
Catalog # 734-1998
Supplier: Corning
Specifications
- Popis:8-channel adapter for use with Costar® vacuum aspirator
- Bal.:1
Specifications
About this item
This is an 8-channel adapter that fits on the Costar® aspirator so that it can be used to remove media and solutions from 8 wells simultaneously on a 96 well plate.