Orders LinkContactUsLinkComponent
Culture system, Corning® E-Cube™
Culture system, Corning® E-Cube™
E-Cube system kit including oxygenator, medium reservoir, multiple access ports, and all required tubing and fittings (without CellCube module), 8500 cm²
Catalog # 734-4053
   Low Manufacturing Impact
Supplier:   Corning
Culture system, Corning® E-Cube™
Catalog # 734-4053
Supplier:  Corning
   Low Manufacturing Impact
Culture system, Corning® E-Cube™
Culture system, Corning® E-Cube™
E-Cube system kit including oxygenator, medium reservoir, multiple access ports, and all required tubing and fittings (without CellCube module), 8500 cm²
Price
143 880,00 Kč
Log in for availability
1 * 1 KIT
Product Details & Documents
The E-Cube™ System is a simple bioreactor with 8500 cm² cell growth area for growing anchorage dependent cells in only a 25,4×35,6 cm footprint. The E-Cube™ System provides a simple method to determine if cells will grow in the CellCube® Module prior to investing in the resources and funding that would be necessary for the larger, automated CellCube® System. Peristaltic pump, CO₂ incubator and medium are not provided.
Includes an oxygenator, medium reservoir, multiple access ports, and all required tubing and fittings.
Cells grow in Corning’'s parallel-plate CellCube® Module on the same treated polystyrene used in Corning culture vessels. The E-Cube™ System kit consists of an oxygenator, medium reservoir, multiple access ports, and all required tubing and fittings; CellCube® Modules must be purchased separately.
Specifications
  • Popis:
    E-Cube system kit including oxygenator, medium reservoir, multiple access ports, and all required tubing and fittings (without CellCube module)
  • Plocha růstu:
    8500 cm²
  • Cat. No.:
    734-4053
  • Certifikát ochrany životního prostředí:
    RECs, ISO 14001 - Kennebunk
  • Environmentally Preferable:
    Low Manufacturing Impact
Accessories
Product Information
  • Catalog #
    Popis
    Materiál
    Availability
    Price
  • 734-4184
    Screw cap, 45 mm, with two stainless steel tubing ports, for E-Cube™ Culture System
    PP
    Log in for availability
    1 * 1 KS - 5 640,00 Kč