Orders LinkContactUsLinkComponent
Staining troughs, Schiefferdecker
Staining troughs, Schiefferdecker
Staining troughs, Schiefferdecker, 70×86×51 mm
Catalog # BRND474410
Supplier:   Brand
Staining troughs, Schiefferdecker
Catalog # BRND474410
Supplier:  Brand
Staining troughs, Schiefferdecker
Staining troughs, Schiefferdecker
Staining troughs, Schiefferdecker, 70×86×51 mm
Price
2 570,00 Kč
Log in for availability
1 * 4 KS
Product Details & Documents
Made of transparent PMP.
This rectangular jar is supplied with a lid and can hold 10 slides or 20 slides back-to-back.
Specifications
  • Popis:
    Staining troughs, Schiefferdecker
  • Š×H×V:
    70×86×51 mm