Order Entry
Czech Republic
ContactUsLinkComponent
 
 
 
(±)-2-Heptanol for synthesis, Sigma-Aldrich®
  8.20619.0100
 :  Merck
 :  
 
 
(±)-2-Heptanol for synthesis, Sigma-Aldrich®
  8.20619.0100
 :  Merck
 :  

 

  • Typ balení:
    Glass bottle
  • Bal.:
    100 mL
  • Sekvence:
    2440

 

 

  • Solubility:
    3.27 g/l (25 °C)
  • Molar Mass:
    116.20 g/mol
  • Boiling Point:
    159 - 161 °C
  • Vapor Pressure:
    1.6 hPa (25 °C)
  • Flash Point:
    59 °C
  • Refractive Index:
    1.421 (20 °C, 589 nm)
  • Explosion Limit:
    0.9 %(V)
  • Density:
    0.82 g/cm3 (20 °C)

 

 

 :
 :13,04682
 :160 °C (1013 hPa)
 :0,82 g/cm³ (20 °C)
 :3,III
 :M-Clarity™ quality level = MQ200
 :208-844-3
 :1987
 :GHS02
 :GHS07
 :Ambient
 :543-49-7
 :C₇H₁₆O
 :MFCD00004587