Chrom ≥99.996% (metals basis), crystalline pieces 2-15 mm (0.08-0.59 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-47-3
:
Formula:Cr
MDL Number:MFCD00010944
Merck Index:12,02288
Boiling Pt:2469 °C (1013 hPa)
Melting Pt:1840 °C
Density:7,2 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-157-5
Related Information
Frequently Bought Together