ContactUsLinkComponent
96 square well, flat bottom plate, Falcon®
Catalog # 734-0984
Supplier:  Corning
   Low Manufacturing Impact
undefined
96 square well, flat bottom plate, Falcon®
Catalog # 734-0984
Supplier:  Corning
   Low Manufacturing Impact
Specifications
  • Poč. jamek:
    96
  • Sterilní:
    +
  • Popis:
    96-square well plate, flat bottom
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Certifikát ochrany životního prostředí:
    ISO 14001 - Bedford
  • Bal.:
    5
  • Objem jamky:
    120 µl
Specifications
About this item
Sterile flat-bottom microplate with lid, made from crystal-grade virgin PS.
  • Sterilised by gamma irradiation
  • Non pyrogenic