Racks for cryoboxes, Nunc™
Catalog # 479-2483
Supplier: Thermo Fisher Scientific
Specifications
- Barevný kód:Stříbrná
- Přihrádky:12
- Š×H×V vnějš.:143,2×140,4×670 mm
- Bal.:1
Specifications
About this item
Aluminium.
- Fits most chest freezers
- Adjustable shelf heights with a maximum of 12 boxes
- Alignment rods secure boxes in position
- Easy to clean, fits most chest freezers