Accessories for Multipette® M4
Dodavatel: EPPENDORF
Certifikáty
O tomto produktu
Frequently Bought Together
Voda, HiPerSolv CHROMANORM® for HPLC
Dodavatel: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Dodavatel: VWR Chemicals
Aceton ≥99%, TECHNICAL
Dodavatel: VWR Chemicals
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals
VWR®, Nitrile Gloves
Dodavatel: VWR Collection
VWR®, Ubrousky čisticí
Dodavatel: VWR Collection
VWR®, Pipety Pasteurovy, High performance
Dodavatel: VWR Collection
VWR® PV4, Spektrofotometr pro oblast viditelného světla
Dodavatel: VWR Collection
Accessories for multi channel pipettes, mechanical, Research® plus
Dodavatel: EPPENDORF
Disposable weighing dishes
Dodavatel: LUDI
Labels, hazardous substance, GHS pictograms
Dodavatel: Brady
Cell culture media, Dulbecco's Modified Eagle Medium (DMEM)
Dodavatel: Biowest
Rukavice, nitrilové, Kimtech™ Purple Nitrile™
Dodavatel: KIMBERLY CLARK
[EN]STOOL LABSIT CASTORS 450-650 MM, ALU 1 * 1 KS
Dodavatel: bimos
Stoppers for conical ground Joints, PE
Dodavatel: Lenz Laborglas GmbH & CO.KG
Accessories for Multipette® M4
Dodavatel: EPPENDORF
Carbendazim ≥97%, Sigma-Aldrich®
Dodavatel: Merck