Dropping bottles, LDPE, Nalgene®
Dodavatel: Thermo Fisher Scientific
Certifikáty
O tomto produktu
LDPE bottle, PE dropping pipette, neoprene bulb.
- Suitable for dispensing most acids, bases and alcohols
- Good temperature resistance
- Easy handling
Dispense at a rate you control, be it one drop at a time or in a stream, with LDPE bottles with dropper assembly.
Specifikace
- Catalog No:
- NALG2416-0125
- Environmentally Preferable:
- Low Manufacturing Impact
- Certifikát ochrany životního prostředí:
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)