VWR® Rota® pH Indicator Papers
Katalogové číslo VWRC109526.0005
Dodavatel: VWR Chemicals
Specifikace
- pH stupnice:1
- pH range:1-10,0
- Popis:Universal Indicator, Rota
- Environmentally Preferable:
- Bal.:5
Specifikace
O tomto produktu
These rolls (4,8 m × 10 mm) have only 1 colour zone and are useful where high precision of measurement is not necessary.