Sampler for liquids, UniSampler
Catalog # 300-1042
Supplier: BURKLE
Specifications
- Capacity:1000 ml
- Popis:UniSampler with hose and sampling bottle
- Environmentally Preferable:
- Bal.:1
Specifications
About this item
Hand operated vacuum pump sampler with a 2,5 metre long PVC hose, the stainless steel weight will sink the end of the hose to the required sampling depth.
- Deep, narrow, and hard to reach spots
- Powerful suction for viscous media, easy to disassemble and clean
- Universal bottle adapter made of PVC for 24, 32, 39 and 50 mm Ø bottle necks and for 100 to 1000 ml sample bottles
Liquid only comes into contact with the suction hose and sample bottle.
Delivery: Supplied in case with handpump, adapter for bottles, sample bottle 1000 ml, PVC suction hose and stainless steel hose weight, 20 seal-it bottle seals.