Orders LinkContactUsLinkComponent
Multi-channel pump heads
Multi-channel pump heads
Certifikáty
Multi-channel pump heads
3 Dostupných variant níže
Product Details & Documents
Multi-channel pump heads, models C4, C8, and C12. Can be used in combination with peristaltic pump drives PD 5001, PD 5101, and PD 5201 with the multi-channel adapter. Up to 12 pump channels can be operated simultaneously. Separate metering to different containers in a single operation.
  • Fast, easy tubing changing
  • Can also be operated with the pump head partly loaded
  • Flow rates can be selected individually for each channel by choosing the tubing dimensions
  • Pulsation reduced with four or eight rollers
  • Cassettes can be replaced even during pumping
Model C4
Eight rollers for low pulsation pumping
Pump head can be fitted with four small cassettes (optional accessories that can be ordered separately)
Partial use of the pump head is possible
Cassettes are easy to install with snap-lock mechanism
Model C8
Four rollers for low pulsation pumping
Pump head can be fitted with eight medium cassettes or four large cassettes
Partial use of the pump head is possible
Medium and large cassettes can be used in combination
Cassettes are easy to install with snap lock mechanism
Model C12
Eight rollers for low pulsation pumping
Pump head can be fitted with 12 small cassettes
Partial use of the pump head is possible
Built in reduction gear enables pumping of very small volumes
Cassettes are easy to install with snap lock mechanism
Product Family Options
Informace o produktu
  • Sekvence
    Dostupnost
    Price
  • 224-1445
    Pump head C 8
    Pump head C 8
    01
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 41 280,00 Kč
    41 280,00 Kč
    1 * 1 KS
       41 280,00 Kč
  • 224-1446
    Pump head C 4
    Pump head C 4
    02
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 28 360,00 Kč
    28 360,00 Kč
    1 * 1 KS
       28 360,00 Kč
  • 224-1447
    Pump head C 12
    Pump head C 12
    03
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 116 340,00 Kč
    116 340,00 Kč
    1 * 1 KS
       116 340,00 Kč
Accessories
Informace o produktu
  • Katalogové číslo
    Dostupnost
    Price
  • Multi-channel adaptor
    Multi-channel adaptor
    224-1349
    Přihlásit se pro dostupnost
    1 * 1 KS - 3 750,00 Kč
Informace o produktu
  • Katalogové číslo
    Dostupnost
    Price
  • Cassette large
    Cassette large
    224-1453
    Přihlásit se pro dostupnost
    1 * 1 KS - 8 160,00 Kč
Informace o produktu
  • Katalogové číslo
    Dostupnost
    Price
  • Cassette medium
    Cassette medium
    224-1452
    Přihlásit se pro dostupnost
    1 * 1 KS - 7 250,00 Kč
Informace o produktu
  • Katalogové číslo
    Dostupnost
    Price
  • Cassette small
    Cassette small
    224-1451
    Přihlásit se pro dostupnost
    1 * 1 KS - 7 200,00 Kč