Orders LinkContactUsLinkComponent
Grinding balls
Grinding balls
02, Grinding balls, 5 mm, Achát
Catalog # 410-9402
Supplier:   LEMKE GERD
Grinding balls
Catalog # 410-9402
Supplier:  LEMKE GERD
Grinding balls
Grinding balls
02, Grinding balls, 5 mm, Achát
Price
28,90 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Agate.
For laboratory ball mills.
Specifications
  • Ø vněj.:
    5 mm
  • Popis:
    Grinding balls
  • Materiál:
    Achát
  • Sekvence:
    02