Stereo zoom microscopes, SMZ-140 and SMZ-143
Supplier: MOTIC
Certificates
About this item
These digital stereo zoom microscopes are suitable for inspection and assembly applications.
- High resolution image
- Anti-fungal properties
- Black and white frosted glass stage inserts
Delivery information: Supplied with a dust cover.
Specifications
- Catalog No:
- 632-8740
- MOTI1100200600834
- MOTI1101002404501
- 630-0154
- 632-9073
- 630-3036
- MOTI1100200600813
- Hlavice:
- 45° sklon, otáčení o 360°
- 45° sklon, otáčení o 360°
- Okuláry:
- WF 10×/20 mm
- WF 10×/20 mm
- Přibližovací skla:
- 1× - 4×
- 1× - 4×
- Pracovní vzdálenost:
- 80 mm
- 80 mm
- Ostření:
- Separated coarse and fine knobs
- Separated coarse and fine knobs
Specifications
Stands
Product Information
Frequently Bought Together