Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Zadání objednávky
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Osvětlení/Lampy
Osvětlení halogenové
Halogen bulbs
Halogen bulbs
Print
Halogen bulbs
852, Halogen bulb, 15 V, 150 W
Catalog # 631-0518
Supplier:
SCHOTT AG Lighting and imaging
Halogen bulbs
Catalog # 631-0518
Supplier:
SCHOTT AG Lighting and imaging
Expand image
Return to Top
Halogen bulbs
852, Halogen bulb, 15 V, 150 W
Specifications
Add to cart
Jump to:
Specifications
Price
750,00 Kč
Log in
for availability
Měrná jednotka
1 * 1 KS
-
+
Add to cart
Return Policy
Product Details & Documents
Specifications
Popis:
Halogen bulb
Sekvence:
852
Napájení:
150 W
Napětí:
15 V
Hypodermic needles, Agani™
#: 613-5386
Supplier: TERUMO