Microplate dispenser, Multidrop® Combi nL
Katalogové číslo 736-0905
Dodavatel: Thermo Fisher Scientific
Specifikace
- Popis:Multidrop® Combi nL
- Environmentally Preferable:Low Manufacturing Impact
- Certifikát ochrany životního prostředí:ISO14001 (Certificate Registration No. EMS 623400)
- Bal.:1
Specifikace
O tomto produktu
Multidrop® Combi nL provides consistent, high throughput dispensing of sub-microlitre volumes into 1536-, 384- and 96-well plates for the biotech and pharmaceutical laboratories. The Combi nL is flexible, dispensing into plates with variable height and allows the user to dispense by rows or columns or individual wells.
- Precise dispensing, with pressurised container and valve technology, across a range 50 nl to 50 µl
- High throughput operation - can be integrated with a range of robotic platforms
- Can dispense all common reagents, diluents, buffers, protein solutions, cells and beads
- Simple operation - graphic display with easy to use menu driven selections or with flexible FILLit PC-software
- Reduced reagent costs - low volume dispensing and minimal dead volumes