Air flow indicator
Katalogové číslo 151-0187
Dodavatel: Dräger
Specifikace
- Popis:Air flow indicator
- Bal.:1 KIT
Specifikace
O tomto produktu
This air flow indicator for special applications can be used in all operations relating to draught detection.
Delivery: Supplied with an aspirator bulb, ten air flow test tubes and two sealing caps.