Automated crude fibre determination, FIBRETHERM®
705-0582
: GERHARDT
- Popis:Automated crude fibre determination, FIBRETHERM®
- Bal.:1
Automated processing of the boiling and filtration steps for the crude fibre, ADF and NDF determination. Ideal for laboratories with high sample throughput.
- Saves time with simultaneous and reproducible analysis of up to 12 samples
- Small footprint and ergonomic design, including pneumatic lift and errors alarm
- Automation of procedure reduces costs and reagents used
- FibreBags provide constant filtration quality
- Safe operation with additions of water, acid and caustic reagents and subsequent filtration that can be programmed
: System is delivered complete.