Multiple horizontal gel electrophoresis system, Owl™ A2-OK
730-0161
- Model:Owl A2-OK
- Popis:Multiple gel system
- Environmentally Preferable:
- Objem pufru:2300 ml
- Velikost gelu Š x D:70×80 mm
- ŠxDxV přístroje:280×375×100 mm
- Bal.:1
Owl™ A2-OK multiple gel system is ideal for teaching labs or labs that need to run multiple gels simultaneously.
- Up to six mini gels can be run
- One teacher can instruct up to 12 students with one system and only one power supply
- The system can screen up to 144 samples at once when used with optional combs (120 with supplied combs)
- EasyCast casting design allows the casting of gels without tape or plugs
- UV transmissible gel trays may be placed directly on the transilluminator for photodocumentaion
: Each unit includes buffer chamber with 3-point levelling base, SuperSafe lid with attached power supply leads, EasyCast gasketed UVT gel trays (6), external casting chambers (2), gel tray holder, and 12 double-sided combs (10-well, 1,0/1,5 mm thick).