Dissolved oxygen probe, StirrOx® G
Dissolved oxygen probe StirrOx® G, StirrOx® G, self-strirring, incl. calibration vessel OxiCal®-ST
Katalogové číslo 664-0510
Dodavatel: WTW
Přejít na:
Product Details & Documents
Self-stirring oxygen probe for simultaneous stirring and measurement.
- Single-handed operation for rapid series measurements
- Constant approach flow for high reproducibility
- Extremely low self-consumption of oxygen
- Temperature compensation
- Membrane leakage monitoring
Informace pro objednávání: Inclusive calibration vessel OxiCal®-ST.
Documents
Sensors for Field and Laboratory.pdf
Další podrobnosti o produktu
Specifikace
- Type:StirrOx® G, self-strirring, incl. calibration vessel OxiCal®-ST
- Popis:Dissolved oxygen probe StirrOx® G
Accessories
Informace o produktu
- Katalogové čísloSekvenceDostupnostPrice
- Electrolyte for galvanic dissolved oxygen sensorsElectrolyte for galvanic dissolved oxygen sensors662-040205Přihlásit se pro dostupnost1 * 50 mL - 2 270,00 Kč
Informace o produktu
- Katalogové čísloDostupnostPrice
- Tripod foot, Sta-ST, for OxiCal®-ST calibration and storage vessel for StirrOx® G oxygen sensorTripod foot, Sta-ST, for OxiCal®-ST calibration and storage vessel for StirrOx® G oxygen sensor662-0407Přihlásit se pro dostupnost1 * 1 KS - 2 270,00 Kč
Informace o produktu
- Katalogové čísloDostupnostPrice
- WP3-ST, exchange membrane heads for StirrOx® GWP3-ST, exchange membrane heads for StirrOx® G662-0409Přihlásit se pro dostupnost1 * 3 KS - 5 130,00 Kč
Informace o produktu
- Katalogové čísloDostupnostPrice
- Fixing cover for Karlsruhe bottle KF 12
Informace o produktu
- Katalogové čísloSekvenceDostupnostPrice
- Calibration and storage vessel for precision calibration in water vapor saturated air, for StirrOx® GCalibration and storage vessel for precision calibration in water vapor saturated air, for StirrOx® GWTWA20534903Přihlásit se pro dostupnost1 * 1 KS - 6 300,00 Kč
Informace o produktu
- Katalogové čísloDostupnostPrice
- Karlsruhe bottles, incl. 60 mm ground stoppers suitable for BOD determinationKarlsruhe bottles, incl. 60 mm ground stoppers suitable for BOD determination715-3254Přihlásit se pro dostupnost1 * 1 KS - 1 510,00 Kč
Informace o produktu
- Katalogové čísloDostupnostPrice
- StirrOx-G Maintenance kit (incl. abrasive foil, 3× exchange membrane heads and electrolyte)StirrOx-G Maintenance kit (incl. abrasive foil, 3× exchange membrane heads and electrolyte)662-0403Přihlásit se pro dostupnost1 * 1 KS - 6 950,00 Kč