Orders LinkContactUsLinkComponent
Dry ice machines, SnowPack
Dry ice machines, SnowPack
Dodavatel:   BURKLE
Dry ice machines, SnowPack
Dodavatel:  BURKLE
Certifikáty
Dry ice machines, SnowPack
Dry ice machines, SnowPack
2 Dostupných variant níže
Product Details & Documents
SnowPack enables the user to obtain a −79 °C dry ice tablet within minutes, without complicated technology and without electricity. Can be installed on any commercial CO₂-bottle.
  • Ideal coolant for transporting
  • Dry ice is odourless and easy to handle
Documents
Product Family Options
Informace o produktu
  • Ø×H
    Dostupnost
    Price
  • 474-3110
    Snowpack® 50 - gas outflow time/tablet (min):~ 0,5
    Snowpack® 50 - gas outflow time/tablet (min):~ 0,5
    50×30 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 8 450,00 Kč
    8 450,00 Kč
    1 * 1 KS
       8 450,00 Kč
  • 474-0057
    Snowpack® 250 - gas outflow time/tablet (min): ~ 2,5
    Snowpack® 250 - gas outflow time/tablet (min): ~ 2,5
    80×60 mm
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 10 380,00 Kč
    10 380,00 Kč
    1 * 1 KS
       10 380,00 Kč