Kjeldahl mineralisation units, Kjeldatherm
Supplier: GERHARDT
Certificates
About this item
Complete units including a integrated heat shield and glass exhaust manifold, insert rack and digestion tubes.
- Insulation without asbestos
- Aluminium heat block with external controller
- Enamelled housing
Manual systems, KB: The handling of the insert rack and manifold are done manually.
Automatic systems, KBL: Kjeldalift motorised lift for the insert rack and manifold.
Scrubber unit, Turbosog: The Turbosog scubber unit condenses and neutralises aggressive acid vapors from the Kjeldahl digestion, including sulphuric acid and other toxic gases. Preseparator has 2 bottles for separation and neutralisation/washing. Water consumption of 30 L/hr. Little maintenance is required as there is no need for activated carbon filters.
Delivery: Manual systems are supplied with PID type electronic digital TR controller providing temperature regulation from 0 to 420 °C and display of actual temperature, with PT 100 probe. Automatic systems are supplied with the programmable TZ control unit, which incorporates a temperature regulator capable of defining and storing nine different temperature and time steps;control init for the Keldahllift, and the Turbosog scrubber unit.