Software, Liqui-Soft® 2.0
Katalogové číslo 612-9837
Dodavatel: HIRSCHMANN
Specifikace
- Popis:Software, liqui-soft® 2.0
- Bal.:1
Specifikace
O tomto produktu
Software for titration, distribution, documentation, and automatic calibration.
- Enables up to four akku-drive®, solarus® or other types of dispensers to be controlled by the PC
- Very user-friendly interface, simple protocol documentation
- Any volumetric measurement instrument can be calibrated and the data required for compliance with DIN and ISO standards are stored in memory
- Simultaneous recording and storage of all measured values
- Compatible with Windows 95, 98, ME, NT, 2000 and XP