VWR®, Hadice vysokoteplotní pro oběhové termostaty
Catalog # 461-0106
Supplier: VWR Collection
Specifications
- Popis:High temperature tubing, for VWR and other circulators
- Bal.:1
Specifications
About this item
PTFE hadice, oplet z nerezové oceli.
Flexible PTFE lined, stainless steel over braided tubing with 6,3 mm NPT quick connectors at both ends. Maximum temperature is 300 °C.
Délka hadice (mm): 930