Crimp top vials and closures, 11 mm
: Agilent
Crimp caps with 11 mm septa
Frequently Bought Together
VWR®, Manual Crimper and Decapper
: VWR Collection
Glass crimp top 0,3 ml microvials for <2 ml samples, level 3 high performance applications, SureSTART™
: Thermo Fisher Scientific
Láhve, čtvercové, úzké nebo široké hrdlo, se závitem
: DWK Life Sciences
Valved Dolomite Light Cup Respirator, FFP3
: Portwest
Dropping bottles, glass, DURAN®
: DWK Life Sciences
Crimp caps with 11 mm septa
: Agilent
Ethanol (untaxed) ≥99.8%, CHROMASOLV™, gradient grade for HPLC, Riedel-de Haën™
: Honeywell Chemicals
Consumables for Agilent Technologies autosamplers
: Trajan Scientific and Medical