Orders LinkContactUsLinkComponent
Non-return valve
Non-return valve
7 mm ID
Katalogové číslo 229-3591
Dodavatel:   Lenz Laborglas
Non-return valve
Katalogové číslo 229-3591
Dodavatel:  Lenz Laborglas
Non-return valve
Non-return valve
7 mm ID
Přejít na:
Price
695,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
DURAN®, borosilicate glass, clear. Prevent pressure loss or backflow. Simple design, cannot be dismantled.
  • Excellent chemical resistance
  • Excellent thermal resistance, autoclavable
  • With tapered ends to ensure that the tube is held firmly
Documents
Specifikace
  • Velikost hadice/trubice:
    7 mm ID