PCR microtubes, 0,2 ml
Dodavatel: EPPENDORF
O tomto produktu
PP.
- 0,2 ml tube has a contamination shield on the hinged lid
- Certified free from DNase, RNase and human genomic DNA
These thin-walled, 0,2 ml PCR tubes are designed for use with all leading thermal cyclers.
Colour: Clear
3 Dostupných variant níže
Product Details & Documents
PP.
- 0,2 ml tube has a contamination shield on the hinged lid
- Certified free from DNase, RNase and human genomic DNA
These thin-walled, 0,2 ml PCR tubes are designed for use with all leading thermal cyclers.
Colour: Clear
Specifications for Product Family
Specifikace
- Catalog No:
- 732-0097
- 732-0098
- EPPE0030124.707
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Certifikát ochrany životního prostředí:
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
Specifikace
Product Family Options
Informace o produktu
- SekvenceBarvaKapacitaBalenéDostupnostPrice
- 732-0097PCR tubes, individual, with attached capPCR tubes, individual, with attached cap01Průhledná0,2 mlPřihlásit se pro dostupnost1 * 1.000 KS4 130,00 KčSpecifikace:
Low Manufacturing Impact More
Specifications
Cat. No.732-0097Environmentally PreferableLow Manufacturing ImpactCertifikát ochrany životního prostředíRenewable Energy Certificate, ISO 14001Low Manufacturing Impact - 732-0098PCR tube strips, 8-tube strips with individually attached caps (960 tubes)PCR tube strips, 8-tube strips with individually attached caps (960 tubes)02Průhledná0,2 mlPřihlásit se pro dostupnost1 * 120 KS5 050,00 KčSpecifikace:
Low Manufacturing Impact More
Specifications
Cat. No.732-0098Environmentally PreferableLow Manufacturing ImpactCertifikát ochrany životního prostředíRenewable Energy Certificate, ISO 14001Low Manufacturing Impact - EPPE0030124.707PCR tubes, Forensic DNA gradePCR tubes, Forensic DNA grade03Průhledná0,2 ml10×50 tubesPřihlásit se pro dostupnost1 * 500 KS2 340,00 KčSpecifikace:
Low Manufacturing Impact More
Specifications
Cat. No.EPPE0030124.707Environmentally PreferableLow Manufacturing ImpactCertifikát ochrany životního prostředíRenewable Energy Certificate, ISO 14001Low Manufacturing Impact
Accessories
Informace o produktu
- Katalogové čísloBalenéDostupnostPrice
- Frame, for work tray (set of 5)