Microcentrifuge tubes, low surface tension, Cliklok™
Katalogové číslo SIMPT330-7LST
Dodavatel: Simport Scientific
Specifikace
- Type:with attached Cliklok™ lid, , low surface tension
- Verze:graduated, conical bottom, non sterile
- Kapacita:1,5 ml
- Barva:Bezbarvá
- Bal.:500
- Max. RCF:20000×g
Specifikace
O tomto produktu
PP, graduated.
- One-piece construction with snug fitting attached cap and reinforced tube bottom for added protection against leakage
- No lubricant (e.g. silicone) needed, thereby eliminating the danger of sample contamination
- Designed to withstand the stress of high speed centrifugation up to 20000×g
Low adhesion surface for optimum sample yield. Cliklok™ seal is especially designed to reduce popping where freezing, boiling, storing, or shipping is required.
Packaging: Packaged in tamper-proof resealable safety-lock bags of 500 tubes. 10 bags per case.