Kleště na kádinky, nerezová ocel
231-4145
: RSG
- Délka:330 mm
- Bal.:1
- Upínací oblast:60 - 150 mm
Stainless steel ¹⁸/₁₀, anti-magnetic. For easy, safe handling of beakers and similar containers. The jaws are wrapped in ceramic fibres to provide thermal insulation and to protect the containers they hold.
- Corrosion resistant
- Excellent thermal resistance, autoclavable