O tomto produktu
Traditional pH indicator test papers, supplied on a 4,8 metre roll, consisting of high quality filter paper impregnated with indicator and mixed indicator solutions then dried and cut to size.
- Roll format are easy to use and most common ranges are available in a refill format
- Quality assurance - tests are checked and calibrated against certified buffer solutions
- Wide and narrow pH ranges available
* no colour scale card supplied
** pack of 3×100 strips
10 Dostupných variant níže
Product Details & Documents
Traditional pH indicator test papers, supplied on a 4,8 metre roll, consisting of high quality filter paper impregnated with indicator and mixed indicator solutions then dried and cut to size.
- Roll format are easy to use and most common ranges are available in a refill format
- Quality assurance - tests are checked and calibrated against certified buffer solutions
- Wide and narrow pH ranges available
* no colour scale card supplied
** pack of 3×100 strips
Documents
Specifications for Product Family
Specifikace
- Catalog No:
- 1.09527.0001
- 1.10232.0001
- 1.09526.0003
- 1.09486.0003
- 1.09489.0003
- 1.09521.0003
- 1.09555.0003
- 1.09557.0003
- 1.09514.0003
- 1.09556.0003
- M-Clarity quality level:
- MQ100
- MQ100
- MQ100
- MQ100
- MQ100
- MQ100
- MQ100
- MQ100
- MQ100
- MQ100
Specifikace
Product Family Options
Informace o produktu
- pH rozsahpH stupniceDostupnostPrice
- 1.09527.0001Universal indicator replacement rolls only*Universal indicator replacement rolls only*1 - 101Přihlásit se pro dostupnost1 * 6 Roll1 770,00 KčSpecifikace:
More
Specifications
Cat. No.1.09527.0001M-Clarity quality levelMQ100 - 1.10232.0001Replacement rolls for pH 1 - 14*Specifikace:
More
Specifications
Cat. No.1.10232.0001M-Clarity quality levelMQ100 - 1.09526.0003Universal indicator, including colour scaleUniversal indicator, including colour scale1 - 101Přihlásit se pro dostupnost1 * 3 Roll1 340,00 KčSpecifikace:
More
Specifications
Cat. No.1.09526.0003M-Clarity quality levelMQ100 - 1.09486.0003Litmus paper blue (pH<7 red / >7 blue)Specifikace:
More
Specifications
Cat. No.1.09486.0003M-Clarity quality levelMQ100 - 1.09489.0003Litmus paper red (pH<7 red / >7 blue)Specifikace:
More
Specifications
Cat. No.1.09489.0003M-Clarity quality levelMQ100 - 1.09521.0003Phenolphthalein paper (pH<8,5 colourless / >8,5 red)Phenolphthalein paper (pH<8,5 colourless / >8,5 red)--Přihlásit se pro dostupnost1 * 3 Roll1 500,00 KčSpecifikace:
More
Specifications
Cat. No.1.09521.0003M-Clarity quality levelMQ100 - 1.09555.0003Special indicatorSpecifikace:
More
Specifications
Cat. No.1.09555.0003M-Clarity quality levelMQ100 - 1.09557.0003Special indicatorSpecifikace:
More
Specifications
Cat. No.1.09557.0003M-Clarity quality levelMQ100 - 1.09514.0003pH-indikační papírek Kongo červeňSpecifikace:
More
Specifications
Cat. No.1.09514.0003M-Clarity quality levelMQ100 - 1.09556.0003Special indicatorSpecifikace:
More
Specifications
Cat. No.1.09556.0003M-Clarity quality levelMQ100