Orders LinkContactUsLinkComponent
Tanks, cylindrical, lightweight
   Sustainable Options Available
Tanks, cylindrical, lightweight
   Sustainable Options Available
Certificates
Tanks, cylindrical, lightweight
5 Options Available Below
Product Details & Documents
LLDPE, translucent with lid. Economical tanks with graduations. With protruding rim (flange). Wall thickness ~2,4 mm. Tank 216-0322 can also be used as an insert for 208 litre steel drums.
  • Good chemical resistance
  • Heat resistant from –100 to +120 °C
  • Conform to the requirements of USP VI and FDA 21 CFR 177.1520
Specifications for Product Family
Specifications
  • Catalog No:
  • 216-0317
  • 216-0318
  • 216-0320
  • 216-0321
  • 216-0322
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Certifikát ochrany životního prostředí:
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
Specifications
Product Family Options
Product Information
  • Popis
    Kapacita
    Ø vněj.×V
    Availability
    Price
  • Lightweight cylindrical tank, graduated, with cover
    19 l
    280×380 mm
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 6 170,00 Kč
    6 170,00 Kč
    1 * 1 KS
       6 170,00 Kč
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    216-0317
    Environmentally Preferable
    Low Manufacturing Impact
    Certifikát ochrany životního prostředí
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • Lightweight cylindrical tank, graduated, with cover
    28 l
    300×460 mm
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 8 660,00 Kč
    8 660,00 Kč
    1 * 1 KS
       8 660,00 Kč
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    216-0318
    Environmentally Preferable
    Low Manufacturing Impact
    Certifikát ochrany životního prostředí
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • Lightweight cylindrical tank, graduated, with cover
    57 l
    330×690 mm
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 8 770,00 Kč
    8 770,00 Kč
    1 * 1 KS
       8 770,00 Kč
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    216-0320
    Environmentally Preferable
    Low Manufacturing Impact
    Certifikát ochrany životního prostředí
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • Lightweight cylindrical tank, graduated, with cover
    114 l
    460×760 mm
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 11 140,00 Kč
    11 140,00 Kč
    1 * 1 KS
       11 140,00 Kč
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    216-0321
    Environmentally Preferable
    Low Manufacturing Impact
    Certifikát ochrany životního prostředí
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • Lightweight cylindrical tank, graduated, with cover
    208 l
    560×910 mm
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 16 160,00 Kč
    16 160,00 Kč
    1 * 1 KS
       16 160,00 Kč
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    216-0322
    Environmentally Preferable
    Low Manufacturing Impact
    Certifikát ochrany životního prostředí
    NAR Certificate of Retirement
      Low Manufacturing Impact
Accessories
Product Information
  • Catalog #
    Sekvence
    Popis
    Availability
    Price
  • 216-2917
    01
    For 19 l cylindrical tanks (NALG11100-0005 and 54100-0005)
    Log in for availability
    1 * 10 KS - 23 390,00 Kč
Product Information
  • Catalog #
    Sekvence
    Popis
    Availability
    Price
  • 216-2924
    04
    For 57 l cylindrical tanks (NALG11100-0015 and 54100-0005)
    Log in for availability
    1 * 10 KS - 20 700,00 Kč
Product Information
  • Catalog #
    Sekvence
    Popis
    Availability
    Price
  • 216-2918
    02
    For 28 l cylindrical tanks (NALG11100-0007 and 54100-0007)
    Log in for availability
    1 * 10 KS - 23 550,00 Kč
Product Information
  • Catalog #
    Sekvence
    Popis
    Availability
    Price
  • 216-2925
    05
    For 113 L cylindrical tanks (NALG11100-0030 and 54100-0030)
    Log in for availability
    1 * 10 KS - 30 970,00 Kč
Product Information
  • Catalog #
    Sekvence
    Popis
    Availability
    Price
  • 216-2926
    06
    For 208 l cylindrical tanks (NALG11100-0055 and 54100-0055)
    Log in for availability
    1 * 10 KS - 31 290,00 Kč